Order Entry
Finland
Orders LinkContactUsLinkComponent
Polysorbate 40

Polysorbate 40

Polysorbate 40
Certificates
Polysorbate 40

4 Options Available Below

Product Details & Documents
Synonyms:  Sorbitan monopalmitate, ethoxylated, Polyoxyethylene sorbitan monopalmitate
  • CAS Number:9005-66-7
  • Formula:C₆₂H₁₂₂O₂₆
  • MDL Number:MFCD00165345
  • Boiling Pt:? 100 °C (1013 hPa)
  • Melting Pt:~ 0,1 °C
  • Density:1.05 g/cm³
  • Flash Pt:>200 °C
  • Storage Temperature:Ambient

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    Clear to hazy light yellow to dark yellow Viscous liquid or paste
  • Infrared spectrum:
    Conforms
  • Acid value:
    ≤3 mg KOH/g
  • Hydroxyl value:
    89 to 105 mg KOH/g
  • Saponification value:
    41 to 55 mg KOH/g
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    25 mL
    Log in for availability
    Log in for availability
    1 * 25 mL
    1 * 25 mL - 21,50 €
    21,50 €
    1 * 25 mL
       21,50 €

  • Glass bottle
    250 mL
    Log in for availability
    Log in for availability
    1 * 250 mL
    1 * 250 mL - 41,30 €
    41,30 €
    1 * 250 mL
       41,30 €

  • Glass bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 95,10 €
    95,10 €
    1 * 1 L
       95,10 €

  • Glass bottle
    2,5 L
    Log in for availability
    Log in for availability
    1 * 2,5 L
    1 * 2,5 L - 180,00 €
    180,00 €
    1 * 2,5 L
       180,00 €