Order Entry
Finland
ContactUsLinkComponent
di-Sodium L(+)-tartrate dihydrate
di-Sodium L(+)-tartrate dihydrate
Catalog # MOLEM76658647
Supplier:  Molekula
CAS Number:  
undefined
di-Sodium L(+)-tartrate dihydrate
Catalog # MOLEM76658647
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

About this item

 :
 :230.08 g/mol
Merck Index:12,08833
Melting Pt:57 °C
Density:1.818 g/cm³ (20 °C)
EINECS:212-773-3
CAS Number:6106-24-7
Formula:NaOOC(CHOH)₂COONa·2H₂O
MDL Number:MFCD00150035