Order Entry
Finland
ContactUsLinkComponent
Paraformaldehyde, EMPROVE® ESSENTIAL DAC, SAFC®
  1.04005.5000
 :  
Paraformaldehyde, EMPROVE® ESSENTIAL DAC, SAFC®
  1.04005.5000
 :  
  • Pack type:
    Plastic bottle for solids
  • Pk:
    5 kg
  • Seq:
    4114
 :
 :13,07096
 :135…136 °C (13 mmHg)
 :123 °C
 :1.46 g/cm³ (20 °C)
 :70 °C
 :2213
 :4.1,III
 :GHS05
 :GHS08
 :GHS07
 :Refrigerator
 :30525-89-4
 :(CH₂O)n
 :MFCD00133991