Polyethylene glycol 600
: Ethylene glycol polymer average Mn 600, PEG 600
- :25322-68-3
- :H(OCH₂CH₂)nOH
- :MFCD00081839
- :600 g/mol
- :? 250 °C (1013 hPa)
- :17…22 °C
- :1.1265 g/cm³
- :252 °C
- :Ambient
- :500-038-2
: Ethylene glycol polymer average Mn 600, PEG 600
- :25322-68-3
- :H(OCH₂CH₂)nOH
- :MFCD00081839
- :600 g/mol
- :? 250 °C (1013 hPa)
- :17…22 °C
- :1.1265 g/cm³
- :252 °C
- :Ambient
- :500-038-2
Documents
- Appearance:Clear colourless to almost colourless Viscous liquid or low melting solid
- Infrared spectrum:Conforms
- Refractive index:1.4660 to 1.4700 (20°C, 589 nm)
- Water:≤2.0 % (K.F.)
- Hydroxyl value:178 to 197 mg KOH/g
Product Family Options
- Pack typePk



