Polyethylene glycol 200, Supelco®
Catalog # 81282-1G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:200 g/mol
Boiling Pt:∼250 °C (1013 hPa)
Melting Pt:–55…–40 °C
Density:1.124 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
Flash Pt:180 °C
Storage Temperature:Ambient
EINECS:500-038-2
CAS Number:25322-68-3
:MFCD00081839
Formula:H(OCH₂CH₂)nOH
MDL Number:MFCD01779596