Order Entry
Finland
ContactUsLinkComponent
J.T.Baker®, Aluminium Standard Solution, 1000 mg/L Al in 0,5 M Nitric Acid (from Al(NO₃)₃)
J.T.Baker®, Aluminium Standard Solution, 1000 mg/L Al in 0,5 M Nitric Acid (from Al(NO₃)₃)
Catalog # 6801.0100
Supplier:  Avantor
CAS Number:  
J.T.Baker®, Aluminium Standard Solution, 1000 mg/L Al in 0,5 M Nitric Acid (from Al(NO₃)₃)
Catalog # 6801.0100
Supplier:  Avantor
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    0,5 M HNO₃
  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Element:
    Aluminium
  • Symbol:
    Al

Specifications

About this item