Bovine serum albumin (BSA)
- Low heavy metals
- CAS No.:9048-46-8
90604-29-8 - Source:Serum
- Species:Bovine
- Storage conditions:Refrigerator
- Protein synonyms:BSA|Fraction V|Cohn Fraction V
- Protein/peptide name:Bovine Serum Albumin
- Colour:Light yellow