Order Entry
Finland
ContactUsLinkComponent
Platinum rhodium alloy (87:13 w%), wire for ISA type R standard grade thermocouple, Ø 0.076 mm (0.003 in)
Platinum rhodium alloy (87:13 w%), wire for ISA type R standard grade thermocouple, Ø 0.076 mm (0.003 in)
Certificates
Platinum rhodium alloy (87:13 w%), wire for ISA type R standard grade thermocouple, Ø 0.076 mm (0.003 in)