Order Entry
Finland
ContactUsLinkComponent
Platinum rhodium alloy (87:13 w%), wire for ISA type R standard grade thermocouple, Ø 0.5 mm (0.02 in)
Platinum rhodium alloy (87:13 w%), wire for ISA type R standard grade thermocouple, Ø 0.5 mm (0.02 in)
Catalog # 13018.BY
CAS Number:  
Platinum rhodium alloy (87:13 w%), wire for ISA type R standard grade thermocouple, Ø 0.5 mm (0.02 in)
Catalog # 13018.BY
CAS Number:  

Specifications

  • Pk:
    100 cm
  • Seq:
    7860

Specifications

About this item

 :
Density:19.595G/CC
Storage Temperature:Ambient
MDL Number:MFCD00198186