Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Nickel Chromium alloy (80:20 w%)
Nickel chromium alloy (80:20 w%), wire, Ø 0.143 mm (0.0056 in)
Nickel chromium alloy (80:20 w%), wire, Ø 0.143 mm (0.0056 in)
Nickel chromium alloy (80:20 w%), wire, Ø 0.143 mm (0.0056 in)
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
Nickel chromium alloy (80:20 w%), wire, Ø 0.143 mm (0.0056 in)
Supplier:
Thermo Fisher Scientific
Certificates
About this item
CAS Number:
11106-97-1
:
MDL Number:
MFCD00673110
:
GHS08
:
GHS07
Storage Temperature:
Ambient
Related Information
Documentation
Certificates
Search by Lot Number
SDS
externalurl.pdf
msds_ALFA013966
Documentation