Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Copper Nickel alloy (55:45 w%)
Copper nickel alloy (55:45 w%), wire, Ø 0.25 mm (0.01 in)
Copper nickel alloy (55:45 w%), wire, Ø 0.25 mm (0.01 in)
Copper nickel alloy (55:45 w%), wire, Ø 0.25 mm (0.01 in)
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
Copper nickel alloy (55:45 w%), wire, Ø 0.25 mm (0.01 in)
Supplier:
Thermo Fisher Scientific
Certificates
About this item
:
MDL Number:
MFCD00801098
Storage Temperature:
Ambient
Related Information
Documentation
Certificates
Search by Lot Number
SDS
msds_ALFA043271
Documentation