Order Entry
Finland
ContactUsLinkComponent
Gold germanium alloy (88:12 w%) ≥99.997% (metals basis), foil, Premion®, Width: 43.8 mm (1.72 in), Thickness: 0.1 mm (0.004 in)
Gold germanium alloy (88:12 w%) ≥99.997% (metals basis), foil, Premion®, Width: 43.8 mm (1.72 in), Thickness: 0.1 mm (0.004 in)
Catalog # 36313.BS
CAS Number:  
Gold germanium alloy (88:12 w%) ≥99.997% (metals basis), foil, Premion®, Width: 43.8 mm (1.72 in), Thickness: 0.1 mm (0.004 in)
Catalog # 36313.BS
CAS Number:  

Specifications

  • Pk:
    10 cm
  • Seq:
    7600

Specifications

About this item

 :
Storage Temperature:Ambient
MDL Number:MFCD02091706