Starch, soluble (from potato), TECHNICAL
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:9005-84-9
:
Formula:(C₆H₁₀O₅)n
MDL Number:MFCD00082026
:162.14 g/mol
Storage Temperature:Ambient
EINECS:232-686-4
Specification Test Results
- Water:Max. 20.5 %
- Fe (Iron):Max. 10 ppm
- Pb (Lead):Max. 0.5 ppm