Gentamicin sulphate
Catalog # PROOICRS0319
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
Merck Index:13,04403
Melting Pt:218…237 °C
EINECS:215-778-9
:GHS08
Storage Temperature:Ambient
CAS Number:1405-41-0
MDL Number:MFCD00270181