Order Entry
Finland
Orders LinkContactUsLinkComponent
L(-)-Mannose ≥99%

L(-)-Mannose ≥99%

L(-)-Mannose ≥99%
Certificates
L(-)-Mannose ≥99%
L(-)-Mannose ≥99%

3 Options Available Below

Product Details & Documents

Purity: 99% CAS No.: 10030-80-5 Molecular Formula: C6H12O6 Formula Weight: 180.16 Melting Point: 129-131° Beilstein Reference: 1724628

Synonyms:  (2R,3R,4S,5S)-2,3,4,5,6-Pentahydroxyhexanal, L-(-)-Mannose
  • CAS Number:10030-80-5
  • Formula:C₆H₁₂O₆
  • MDL Number:MFCD00136021
  • Molecular Weight:180.16 g/mol
  • Boiling Pt:376 °C (1013 hPa)
  • Melting Pt:129…131 °C
  • Storage Temperature:Ambient
  • EINECS:233-080-2
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    0,25 g
    Log in for availability
    Log in for availability
    1 * 0,25 g
    1 * 0,25 g - 99,40 €
    99,40 €
    1 * 0,25 g
       99,40 €

  • Glass bottle for solids
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 296,00 €
    296,00 €
    1 * 1 g
       296,00 €

  • Glass bottle for solids
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 1 160,00 €
    1 160,00 €
    1 * 5 g
       1 160,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.