
TRITON™ X-114 (Polyethylene glycol tert-octylphenyl ether) 10% in aqueous solution, Millipore®
Catalog # 648468-50
Supplier: Merck Millipore (Calbiochem)
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:50 mL
- Seq:2370
Specifications
About this item
:
:GHS05
Storage Temperature:Ambient
CAS Number:9036-19-5
MDL Number:MFCD00128254