Order Entry
Finland
Orders LinkContactUsLinkComponent
D-(+)-Glucose

D-(+)-Glucose

Plastic bottle for solids, 500 g
Catalog # MOLE58499289-500G
Marketsource
Supplier:   Molekula
D-(+)-Glucose
Catalog # MOLE58499289-500G
Supplier:  Molekula
Marketsource
CAS Number:  50-99-7
D-(+)-Glucose
D-(+)-Glucose

Plastic bottle for solids, 500 g

CAS Number:  50-99-7
Price
76,00 €
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  Corn sugar, ᴅ-(+)-Glucose, Glucose, D-(+)-Dextrose, (2R,3S,4R,5R)-2,3,4,5,6-Pentahydroxyhexanal, Glucolin, Dextrose, D-Glucose
  • CAS Number:50-99-7
  • Formula:C₆H₁₂O₆
  • MDL Number:MFCD00148912
  • Molecular Weight:180.16 g/mol
  • Merck Index:14,04459
  • Melting Pt:146 °C
  • Density:1.54 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-075-1
Specifications

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    500 g

Specifications