Heat transfer liquids, Kryo 60
Supplier: LAUDA
Certificates
About this item
These transfer fluids allow highly accurate thermostating, even at extreme temperatures.
- Durability
- Simple and safe handling
- Reliability, suitable for long-term operation
Recommended temperature range: –60 to +60 °C (open system).
* Denotes silicone oil