Heat transfer liquids, Kryo 95
Supplier: LAUDA
Certificates
About this item
Open systems are atmospherically open, as semi-open systems are referred to open baths with lids. The thermally active heat transfer medium in closed cold oil superimposed circuits has no direct contact with ambient air.
- Boiling point: >190 °C
- Density: 855 kg / m³
- Solvents used are petrol, acetone, alcohols
Recommended temperature range: –95 to +60 °C (open system) and –95 to +160 °C (closed system).