Order Entry
Finland
ContactUsLinkComponent
Acrylamide : Bisacrylamide (19:1)
Acrylamide : Bisacrylamide (19:1)
Catalog # SHETIB70023
Supplier:  IBI SCIENTIFIC
CAS Number:  
Acrylamide : Bisacrylamide (19:1)
Catalog # SHETIB70023
Supplier:  IBI SCIENTIFIC
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    200 g
  • Seq:
    3505

Specifications

About this item

 :
Storage Temperature:Refrigerator
UN:3426
ADR:6.1,III
 :GHS06
 :GHS08
 :79-06-1
MDL Number:MFCD00080848