Order Entry
Finland
ContactUsLinkComponent
tert-Butyl-4-methoxyphenol (mixture of isomers) ≥98.5% Ph. Eur., NF, E320, SAFC®
Ärsyttävä
tert-Butyl-4-methoxyphenol (mixture of isomers) ≥98.5% Ph. Eur., NF, E320, SAFC®
Catalog # 8.17023.9025
CAS-numero:  
Ärsyttävä
tert-Butyl-4-methoxyphenol (mixture of isomers) ≥98.5% Ph. Eur., NF, E320, SAFC®
Catalog # 8.17023.9025
CAS-numero:  

Tekniset tiedot

  • Pk:
    25 kg
  • Seq:
    4283

Tekniset tiedot

Määrittelytestin tulokset

  • Assay (HPLC, butylated hydroxyanisole):
    ≥ 98.5 %
  • Assay (HPLC, 2-tert-butyl-4-methoxyphenol):
    ≥ 90.0 %
  • Assay (HPLC, 3-tert-butyl-4-methoxyphenol):
    ≤ 10.0 %
  • Identity (TLC):
    passes test
  • Identity (coloring 1):
    passes test
  • Identity (coloring 2):
    passes test
  • Identity (HPLC):
    passes test
  • Identity (IR-spectrum):
    passes test
  • Appearance:
    White, yellowish or slightly pinkish crystalline powder.
  • Appearance of solution (100 g/l, ethanol 96 %):
    Clear and not more intensely colored than intensity 5 of the range of reference solutions of the most appropriate coloracc. to Ph. Eur.
  • Melting range:
    48 - 63 °C
  • Absorption maximum λ 1 (Ethanol):
    289 - 292 nm
  • Absorption maximum λ 2 (Ethanol):
    227 - 230 nm
  • Spec. Absorptivity A 1%/1cm (λ1max; 0.02 g/l; ethanol):
    190 - 210
  • Spec. Absorptivity A 1%/1cm (λ2max; 0.02 g/l; ethanol):
    326 - 345
  • Heavy metals (as Pb):
    ≤ 10 ppm
  • As (Arsenic):
    ≤ 3 ppm
  • Hg (Mercury):
    ≤ 1 ppm
  • Pb (Lead):
    ≤ 2 ppm
  • Related substances (TLC) (3-tert-Butyl-4-methoxyphenol):
    ≤ 10 %
  • Related substances (TLC) (Hydroquinone):
    ≤ 0.2 %
  • Related substances (TLC) (any other impurity):
    ≤ 0.5 %
  • Residual solvents (ICH Q3C):
    excluded by manufacturing process
  • Sulfated ash (600 °C):
    ≤ 0.01 %

Määrittelytestin tulokset

Tietoja tästä tuotteesta

 :
 :180.25 g/mol
Merck-indeksi:12,01582
Kiehumispiste:264 °C (1013 hPa)
Sulamispiste:48…63 °C
Tiheys:1.05 g/cm³ (20 °C)
Leimahduspiste:156 °C
Varastointilämpötila:Ambient
EINECS:204-442-7
 :GHS07
CAS-numero:25013-16-5
Kaava:C₁₁H₁₆O₂
MDL-numero:MFCD01779059