

Ethyl propenyl ether cis- and trans-mixture ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:928-55-2
:
Formula:C₅H₁₀O
MDL Number:MFCD00015184
:86.13 g/mol
Boiling Pt:69…71 °C
Density:0.778
Flash Pt:20 °C (68 °F)
Storage Temperature:Ambient
EINECS:213-176-0
UN:3271
ADR:3,II
:GHS02
:GHS07
Related Information
Frequently Bought Together