Order Entry
Finland
Orders LinkContactUsLinkComponent
Nickel

Nickel

1
Catalog # PROONIST-772A
Health
Nickel
Catalog # PROONIST-772A
CAS Number:  7440-02-0
CAS Number:  7440-02-0
Price
1 300,00 €
Log in for availability
1 * 1 Items
Product Details & Documents
  • CAS Number:7440-02-0
  • Formula:Ni
  • MDL Number:MFCD00011137
  • Molecular Weight:58.69 g/mol
  • Merck Index:12,06582
  • Boiling Pt:3007 °C (1013 hPa)
  • Melting Pt:1453 °C
  • Density:8.9 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:231-111-4
  • UN:3089
  • ADR:4.1,III
Specifications

Specifications

  • Pk:
    1

Specifications