Niobium ≥99.95% (metals basis), ingot/button, Ø ∼25.4 mm (1.0 in), Thickness: 11 mm (0.45 in)
Catalog # 45113.KF
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1
- Seq:0001
Specifications
About this item
CAS Number:7440-03-1
:
Formula:Nb
MDL Number:MFCD00011126
:92.91 g/mol
Boiling Pt:3700 °C (1013 hPa)
Melting Pt:2468 °C
Density:8.57 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-113-5