Order Entry
Finland
ContactUsLinkComponent
Niobium, gauze woven from wire 40 mesh, Ø 0.178 mm (0.007 in)
Niobium, gauze woven from wire 40 mesh, Ø 0.178 mm (0.007 in)
Catalog # 40916.FL
CAS Number:  
Niobium, gauze woven from wire 40 mesh, Ø 0.178 mm (0.007 in)
Catalog # 40916.FL
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    0001
  • Pk Info:
    75×75 mm

Specifications

About this item

CAS Number:7440-03-1
 :
Formula:Nb
MDL Number:MFCD00011126
 :92.91 g/mol
Boiling Pt:3700 °C (1013 hPa)
Melting Pt:2468 °C
Density:8.57 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-113-5