Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Titanium
Titanium ≥93%, powder <20 µm
Titanium ≥93%, powder <20 µm
:
Thermo Fisher Scientific
Titanium ≥93%, powder <20 µm
:
Thermo Fisher Scientific
:
7440-32-6
:
:
Ti
:
MFCD00011264
:
47.88 g/mol
:
3277 °C (1013 hPa)
:
1677 °C
:
4.5 g/cm³ (20 °C)
:
Ambient
:
231-142-3
:
1352
:
4.1,II
:
GHS02
msds_ALFA000681
Thermo Scientific Acroseal 25 Years