Order Entry
Finland
ContactUsLinkComponent
Aluminium
Aluminium
Catalog # PROOLGC2612
CAS Number:  
Aluminium
Catalog # PROOLGC2612
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :26.98 g/mol
Merck Index:13,00321
Boiling Pt:2327 °C (1013 hPa)
Melting Pt:660 °C
Density:2.7 g/cm³ (20 °C)
EINECS:231-072-3
CAS Number:7429-90-5
Formula:Al
MDL Number:MFCD00134029