Order Entry
Finland
ContactUsLinkComponent
Bond tack cloth
Bond tack cloth
Catalog # CONTBS20CB-12
Supplier:  CONTEC
undefined
Bond tack cloth
Catalog # CONTBS20CB-12
Supplier:  CONTEC
Specifications
  • Packed:
    100/bag, 5 bags/case
  • Description:
    Tack cloth
  • L×W:
    229×305 mm
  • Pk:
    500
Specifications
About this item
These polyester cloths are ideal for the surface preparation prior to painting. Also suitable for particle removal in printed circuit board manufacturing and for eliminating solvents in some applications.
  • Custom blended tack formula for optimal adhesive texture, transfer resistance, heat resistance and anti-static performance
  • Sealed edge technology minimises particles and fibres
  • "Napped" surface easily removes dirt particles
  • Water-based tack is applied at high heat under pressure