Gel extraction kit, GeneJET
GeneJET Gel Extraction Kit is designed for rapid and efficient purification of DNA fragments from standard or low-melting point agarose gels run in either TAE or TBE buffer.
- Highly efficient up to 95% recoveries in the range of 100 bp to 10 kb
- Procedure takes only 15 minutes
- Spin columns are capped and assembled with collection tubes
The kit utilises a proprietary silica-based membrane technology in the form of a convenient spin column. The kit can be used to purify DNA fragments from 25 bp to 20 kb in size with recovery rates up to 95%. Each GeneJET purification column has a binding capacity of up to 25 µg of DNA and can process up to 1 g of agarose gel. The entire procedure takes just 15 minutes, and the isolated DNA is ready to use in all common downstream applications, including ligation, restriction digestion, PCR, sequencing, and labeling.
- Method / Format:Column (silica-based)
- Prep size:Mini
- Refine target molecule:Amplified/modified DNA
- Sample type:Gel matrix
- Target molecule:DNA
- Sample size:50 preps
- Description:GeneJET Gel Extraction Kit