Cathepsin K drug discovery kit
Catalog # ENZOBMLAK4300001
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:1 KIT
- Description:Cathepsin K drug discovery kit
- Seq:02
Specifications
About this item
The Cathepsin K Drug Discovery Kit is a complete assay system designed to screen inhibitors of cathepsin K. Using a convenient microplate format, cathepsin K activity is monitored fluorimetrically using the substrate provided in the kit.
Ordering information: Quantity: 96 assays Contents: Cathepsin K (human) (recombinant), AMC substrate, AMC calibration standard, inhibitor, assay buffer, ½-volume white microplate.