Order Entry
Finland
ContactUsLinkComponent
Rat recombinant Biliverdin Reductase (from E. coli)
Rat recombinant Biliverdin Reductase (from E. coli)
Supplier:  ENZO LIFE SCIENCES
Certificates
Rat recombinant Biliverdin Reductase (from E. coli)
Supplier:  ENZO LIFE SCIENCES

Specifications

  • Enzyme type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Rat
  • Enzyme name:
    Biliverdin reductase
  • Purity:
    Purified

Specifications