Order Entry
Finland
ContactUsLinkComponent
Human recombinant TNF alpha (1) (from E. coli)
Human recombinant TNF alpha (1) (from E. coli)
Catalog # ENZOADI908066010
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Human recombinant TNF alpha (1) (from E. coli)
Catalog # ENZOADI908066010
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    10 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Seq:
    02
  • Protein/peptide name:
    TNF alpha (1)
  • Purity:
    >98% (SDS-PAGE; RP-HPLC)
  • Formulation:
    Lyophilized. Sterile filtered liquid, pH 7,2, containing 20 mM PBS and 10 mM sodium chloride.
  • Purification:
    Purified by multi-step chromatography.

Specifications

About this item

Produced in E.coli.