Order Entry
Finland
ContactUsLinkComponent
Orange 552 dUTP (lyophilised)
Orange 552 dUTP (lyophilised)
Catalog # ENZOENZ42842L0050
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
undefined
Orange 552 dUTP (lyophilised)
Catalog # ENZOENZ42842L0050
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    50 Nmo
  • Description:
    Orange 552 dUTP (lyophilised)
  • Seq:
    02
  • Purity:
    ≥93% (HPLC)
  • Formulation:
    Lyophilised

Specifications

About this item