Order Entry
Finland
ContactUsLinkComponent
Red 580 dUTP
Red 580 dUTP
Catalog # ENZOENZ42844
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Red 580 dUTP
Catalog # ENZOENZ42844
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    25 Nmo
  • Description:
    Red 580 dUTP
  • Seq:
    02
  • Purity:
    ≥93% (HPLC)
  • Concentration:
    1 mM
  • Formulation:
    Liquid. Solution in water.

Specifications

About this item

Red 580 [5-ROX] dUTP can replace TTP in reactions in which it serves as a substrate for E. coli DNA polymerase (holoenzyme and Klenow fragment), T4 and Taq DNA polymerases, reverse transcriptase (from AMV and M-MuLV) and terminal transferase. Fluorescently labelled probes can be prepared with this fluorescent nucleotide by a variety of methods including nick translation, random prime labelling, cDNA labelling and 3’-end labelling. Probes generated by these methods are suitable for use for the identification of specific sequences by in situ hybridization procedures on fixed cells and tissues by direct fluorescence detection. Red 580 dUTP can also be used for multicolor fluorescence labelling.