Order Entry
Finland
ContactUsLinkComponent
ZnAF-2.tetrahydrochloride
ZnAF-2.tetrahydrochloride
Catalog # ENZOALX620072M001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
ZnAF-2.tetrahydrochloride
Catalog # ENZOALX620072M001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Description:
    ZnAF-2 tetrahydrochloride
  • Seq:
    02
  • Purity:
    ≥98% (HPLC)
  • Concentration:
    ~5 mM
  • Formulation:
    Liquid. In 0,28 ml DMSO.

Specifications

About this item

Fluorescent probe for Zn2+

Non cell permeable fluorescent reagent (Ex(max): 492nm; Em(max): 514nm) for the detection of low concentration of zinc ion due to its strong affinity to zinc ion (dissociation constant: 2.7nM). The sample zinc ion can be specifically detected. Low background fluorescense supersensitizes the visualization for in vivo sample zinc ion. Also available as cell permeable derivative (Prod. No. ALX-620-076). ZnAF-2 conjugates TPEN (Prod. No. ALX-450-011) affinities with fluorescein and enhances the specificity for Zn2+.