JC virus BioProbe®
Catalog # ENZOENZ40847
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:2 µG
- Description:JC virus BioProbe®
- Seq:02
- Concentration:20 µg/ml
- Formulation:Liquid. In 10 mM TRIS HCl, pH 7,5, containing 1 mM EDTA.
- Size:2 µg
Specifications
About this item
The JC Virus BioProbe® labeled probe is prepared by nick translation of a clone of the entire JC virus genome. JC virus, a member of the papovavirus family, usually causes subclinical infections. However, in immunocompromised individuals it can cause a fatal demyelinating brain disease, progressive multifocal leukoencephalopathy. JC virus is also neurooncogenic in primates. Fragment size range: 100-1000 base pairs (as estimated by agarose gel electrophoresis).