Order Entry
Finland
ContactUsLinkComponent
Adenovirus BioProbe®
Adenovirus BioProbe®
  ENZOENZ40834
 :  
Adenovirus BioProbe®
  ENZOENZ40834
 :  

 

  • Pk:
    2 µG
  • Description:
    Adenovirus BioProbe®
  • Seq:
    02
  • Concentration:
    20 µg/ml
  • Formulation:
    Liquid. In 10 mM TRIS HCl, pH 7,5, containing 1 mM EDTA.
  • Size:
    2 µg

 

 

The Adenovirus BioProbe® labeled probe is prepared by nick translation of cloned fragments of the adenovirus 5 genome. The fragments constitute a total of 20 kb of DNA, approximately 56% of the genome. The probe is specific for adenovirus and, in addition to hybridization to adenovirus 5 sequences, the probe will cross hybridize with adenovirus types 4, 7, 11, 20, 40 and 41 due to their considerable sequence homology. Fragment size range: 100-1000 base pairs (as estimated by agarose gel electrophoresis).