Order Entry
Finland
ContactUsLinkComponent
 
TIMP-1 (human) ELISA kit
Catalog # ENZOENZKIT1470001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
TIMP-1 (human) ELISA kit
Catalog # ENZOENZKIT1470001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Primary antibody reactivity:
    Human
  • Target protein:
    TIMP-1
  • Description:
    TIMP-1 (Human) ELISA kit, grow cells
  • Seq:
    01
  • Sensitivity:
    ≤30 pg/ml (0,049 - 12 ng/ml)

Specifications

About this item

Highly sensitive TIMP-1 ELISA kit enabling detection of TIMP-1 in serum, plasma, saliva, urine and tissue culture media in just 2 hours.

  • Highly sensitive measurement of TIMP-1, detecting as little as 30 pg/ml
  • Negligible cross-reactivity with similar proteins (TIMP-2, TIMP-3, TIMP-4, MMP-1, MMP-2, MMP-3, MMP-7, MMP-8, MMP-9, MMP-13)
  • High throughput format with results in two hours for up to 39 samples in duplicate
  • Fully quantitative results that surpass semi-quantitative Western blot analysis

For the quantitative determination of TIMP-1 in human serum, plasma, saliva, urine and tissue culture media. The TIMP-1 ELISA kit is a colorimetric immunometric immunoassay kit with results in two hours.

Ordering information: Contains Microtiter plate, Assay Buffer 13, Standard, Wash Buffer Concentrate, Antibody, Antibody Diluent, Conjugate, Substrate, and Stop Solution.