Transferrin ELISA kit, Enzo Life Sciences
Catalog # ENZOENZKIT1430001
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:1 KIT
- Assay Type:Competitive
- Target protein:Transferrin
- Description:Transferrin ELISA kit, grow cells
- Seq:01
- Sensitivity:4,6 ng/ml (4,9 - 5000 ng/ml)
Specifications
About this item
Highly sensitive Transferrin ELISA kit enabling detection of transferrin in serum and plasma in just 2 hours.
- Sensitive measurement of transferrin, detecting as little as 4,6 ng/ml
- Negligible cross reactivity with similar proteins
- High throughput format with results in 2 hours for up to 40 samples in duplicate
- Fully quantitative results that surpass semi-quantitative Western blot analysis
For the quantitative determination of transferrin in human serum, human plasma and canine serum. The Transferrin ELISA kit is a colorimetric competitive immunoassay kit with results in 2 hours.
Ordering information: Contains Microtiter Plate, Assay Buffer 13, Standard, Wash Buffer Concentrate, Antibody, Antibody Diluent, Tracer, Conjugate, Substrate, and Stop Solution.