Order Entry
Finland
ContactUsLinkComponent
Methotrexate ELISA kit
Methotrexate ELISA kit
Catalog # ENZOENZKIT1420001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Methotrexate ELISA kit
Catalog # ENZOENZKIT1420001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Assay Type:
    Competitive
  • Target protein:
    Methrotrexate
  • Description:
    Methotrexate ELISA kit, grow cells
  • Seq:
    01
  • Sensitivity:
    0,087 ng/ml (0,13 - 1000 ng/ml)

Specifications

About this item

Ultra sensitive Methotrexate ELISA kit enabling detection of Methotrexate in serum, plasma, and urine in just 1,5 hours.

  • Sensitive measurement of methotrexate, detecting as little as 0,087 ng/ml
  • Broad dynamic range suitable for a large variety of samples
  • High throughput format with results in 1,5 hours for up to 38 samples in duplicate
  • Fully quantitative results that surpass semi-quantitative Western blot analysis

For the quantitative determination of methotrexate in serum, plasma and urine in human, mouse, and rat samples. The Methotrexate ELISA kit is a colorimetric competitive immunoassay kit with results in 1,5 hours.

Ordering information: Contains Microtiter Plate, Assay Buffer 13, Standard, Wash Buffer Concentrate, Antibody, Antibody Diluent, Conjugate, Substrate, and Stop Solution.