Order Entry
Finland
ContactUsLinkComponent
 
5-Hydroxymethylcytosine DNA ELISA Kit, Grow Cells
  ENZOADI9002250001
 :  
5-Hydroxymethylcytosine DNA ELISA Kit, Grow Cells
  ENZOADI9002250001
 :  

 

  • Pk:
    1 KIT
  • Target protein:
    5-Hydroxymethylcytosine DNA
  • Description:
    5-Hydroxymethylcytosine DNA ELISA kit, grow cells
  • Seq:
    02
  • Sensitivity:
    <0,02% 5-hydroxymethylcytosine DNA per 100 ng input DNA (range 0,03 - 0,55% (100 ng/µl))

 

 

Highly sensitive, rapid ELISA kit to quantify 5-hydroxymethylcytosine in DNA with results that closely correlate to mass spectrometry analysis.

  • Rapid: Fully quantitative results in less than three hours
  • Convenient: User-friendly alternative to mass spectrometry
  • Simple: Efficient and well-established protocol
  • High-throughput: 96-well format ideal for global detection, tissue-specific quantitation, high-throughput compound screening, and more

For the quantitative determination of species independent 5-hydroxymethylcytosine DNA in a variety of DNA samples.

The 5-Hydroxymethylcytosine DNA ELISA kit is a complete, colorimetric, immunometric immunoassay kit for the quantitative determination of 5-hydroxymethylcytosine in DNA  with results in just three hours.

 : Contains Microtiter Plate, Coating Buffer, ELISA Buffer, Primary Antibody, Antibody-conjugate,  HRP Developer, Controls.