Order Entry
Finland
ContactUsLinkComponent
DNA damage ELISA Kit
DNA damage ELISA Kit
Catalog # ENZOADIEKS350
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
DNA damage ELISA Kit
Catalog # ENZOADIEKS350
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Assay Type:
    Competitive
  • Description:
    DNA damage ELISA Kit, Enzo Life Sciences
  • Seq:
    02
  • Sensitivity:
    0,59 ng/ml (range 1,875 - 60 ng/ml)

Specifications

About this item

Rapid DNA damage ELISA kit for cancer, apoptosis and oxidative stress.

For the measurement of DNA Damage (8-OHdG) in serum, saliva, and urine from any species. Cited sample types include plasma, seminal fluids, cell lysates, culture supernatants and DNA extracts.

The DNA Damage EIA kit is a colorimetric competitive enzyme immunoassay kit with results in < 2,5 hours. This kit has been validated in-house in a variety of sample matrices and is supplied in a convenient 96 well plate format. Protocol (xls) available upon request.

Ordering information: Contains Microtiter plate, Standard, Sample diluent, Wash buffer concentrate, Antibody, Antibody diluent, Conjugate, Conjugate diluent, TMB Substrate, Stop solution 2.