Order Entry
Finland
ContactUsLinkComponent
β-Catenin ELISA kit
β-Catenin ELISA kit
Catalog # ENZOADI900135
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
β-Catenin ELISA kit
Catalog # ENZOADI900135
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Primary antibody reactivity:
    Human
    Rat
    Mouse
  • Target protein:
    β-Catenin
  • Description:
    β-Catenin ELISA kit
  • Seq:
    02
  • Sensitivity:
    26,8 pg/ml (range 125 - 8,000 pg/ml)

Specifications

About this item

For the quantitative determination of β-Catenin in cell lysates of human, mouse, and rat origin.

The β-Catenin EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in 3 hours that measures pan β-Catenin. Absorbance is read at 450 nm. This kit has been validated in-house with human, mouse, and rat cell lysates and correlated Western blot. The β-Catenin EIA kit detects as little 27 pg/ml β-catenin surpassing semi-quantitative Western blot analysis and provides simple liquid color-coded reagents that reduce laboratory error.

Ordering information: Contains Microtiter plate, Conjugate, Antibody, Assay buffer 13, RIPA Cell lysis buffer 2, Wash buffer concentrate, Standard, TMB Substrate, Stop solution 2.