Order Entry
Finland
ContactUsLinkComponent
Estriol ELISA kit
Estriol ELISA kit
Catalog # ENZOADI900100
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Estriol ELISA kit
Catalog # ENZOADI900100
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Assay Type:
    Competitive
  • Target protein:
    Estriol
  • Description:
    Estriol ELISA kit
  • Seq:
    02
  • Sensitivity:
    59,6 pg/ml (range 122 - 500,000 pg/ml)

Specifications

About this item

Certified for a large variety of sample types for steroid and hormone research.

For the quantitative determination of Estriol in culture supernatants, plasma, serum, saliva, and urine from any species.

The Estriol EIA kit is a colorimetric competitive enzyme immunoassay kit with results in 3 hours. Absorbance is read at 405 nm. This kit has a broad dynamic range suitable for a wide variety of sample matrices and the ability to <60 pg/mL estriol.

Ordering information: Contains GxR IgG Microtiter plate, Assay buffer concentrate, Conjugate, Antibody, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution.