Cortisol ELISA kit
Supplier: ENZO LIFE SCIENCES
Certificates
About this item
The Cortisol EIA kit is a colorimetric competitive enzyme immunoassay kit for the quantitative determination of cortisol in culture supernatants, plasma, serum, saliva, and urine from any species.
- Trusted - highly cited in peer reviewed literature
- Reproducible results day-after-day and lot-after-lot
- Highly sensitive, detecting low levels of cortisol in biological matrices
- Pre-coated plates and liquid colour coded reagents save time and reduce errors
Absorbance is read at 405 nm. The broad dynamic range allows accurate measurement of cortisol levels in a variety of sample matrices.
Ordering information: Contains GxM IgG microtiter plate, conjugate, antibody, assay buffer, wash buffer concentrate, standard, pNpp substrate, stop solution, steroid displacement reagent.
Specifications
- Assay Type:Competitive
- Storage conditions:4 °C (long term)
- Sensitivity:56,72 pg/ml (range 156 - 10000 pg/ml)