Order Entry
Finland
ContactUsLinkComponent
PGE1 ELISA kit
PGE1 ELISA kit
Catalog # ENZOADI900005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
PGE1 ELISA kit
Catalog # ENZOADI900005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Assay Type:
    Competitive
  • Target protein:
    PGE1
  • Description:
    PGE1 ELISA kit
  • Seq:
    02
  • Sensitivity:
    5,58pg/mL (range 4,88 - 5,000pg/mL)

Specifications

About this item

For the quantitative determination of PGE1 in culture supernatants, plasma, serum, saliva, and urine from any species. Cited sample types include simulated interstitial lung fluid.

The PGE1 EIA kit is a colorimetric competitive enzyme immunoassay kit with results in < 3 hours. Absorbance is read at 405nm. Assay up to 38 samples in duplicate per kit with excellent detection sensitivity.

Ordering information: Contains DxS IgG Microtiter plate, Conjugate, Antibody, Assay buffer, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution. Alternative size available: 89141-274 (5x96 wells)