Order Entry
Finland
ContactUsLinkComponent
cGMP ELISA kit
cGMP ELISA kit
Catalog # ENZOADI901013
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
cGMP ELISA kit
Catalog # ENZOADI901013
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Assay Type:
    Competitive
  • Target protein:
    cGMP
  • Description:
    cGMP ELISA kit
  • Seq:
    02
  • Sensitivity:
    (non-acetylated) 0,37 pmol/ml; (acetylated) 0,088 pmol/ml (range (non-acetylated) 0,16 - 500 pmol/ml; (acetylated) 0,16 - 100 pmol/ml)

Specifications

About this item

Most sensitive (0,088 pmol/ml) colorimetric kit  for extracellular cGMP quantification.

For the quantitative determination of cGMP in culture supernatants, plasma, serum, saliva, and urine from any species.

This is a colorimetric competitive immunoassay kit for the quantitative determination of extracellular cGMP. Sensitivity is increased by acetylation (reagents included). Absorbance is read at 405 nm.The ready-to-use liquid color-coded reagents provided reduce error.

Ordering information: Contains GxR Microtiter plate, Conjugate, Antibody, Assay buffer 2 concentrate, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution, Acetic anhydride, Triethylamine. Alternative size available: 89141-066 (1×96 wells).