Order Entry
Finland
ContactUsLinkComponent
17β-Estradiol high sensitivity ELISA kit
17β-Estradiol high sensitivity ELISA kit
Supplier:  ENZO LIFE SCIENCES
Certificates
undefined
17β-Estradiol high sensitivity ELISA kit
Supplier:  ENZO LIFE SCIENCES

Specifications

  • Assay Type:
    Competitive
  • Target protein:
    17β-Estradiol
  • Storage conditions:
    4 °C (long term), –20 °C (for standards)
  • Sensitivity:
    14,0 pg/ml (range 15,6 - 1000 pg/ml)

Specifications