Order Entry
Finland
ContactUsLinkComponent
HSF1 ELISA Kits
HSF1 ELISA Kits
Catalog # ENZOADI900198
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
HSF1 ELISA Kits
Catalog # ENZOADI900198
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Description:
    HSF1 ELISA kit
  • Seq:
    02
  • Storage conditions:
    4 °C (long term), –20 °C (for standards and microtiter plate)
  • Sensitivity:
    35 pg/ml (Range 0,39 - 12,5 ng/ml)

Specifications

About this item

The HSF1 EIA kit is a complete kit for the quantitative determination of HSF1 in cell lysates and tissue extracts from human and mouse origins. This is a Sensitive ELISA kit for this key transcription factor for heat shock protein (chaperone) research.

  • Use less precious sample compared to Western blot
  • Ready to use liquid color-coded reagents and pre-coated plate reduces error

For the quantitative determination of human and mouse HSF1 in cell lysates and tissue.

Delivery: Supplied with antibody, assay buffer, conjugate, extraction reagent #1, microtiter plate, standard, stop solution, TMB substrate, wash buffer concentrate.