Order Entry
Finland
ContactUsLinkComponent
IL-2 (human), ELISA kit
IL-2 (human), ELISA kit
Catalog # ENZOADI900118A
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
IL-2 (human), ELISA kit
Catalog # ENZOADI900118A
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Human
  • Target protein:
    IL-2
  • Description:
    IL-2 (Human), ELISA kit
  • Seq:
    02
  • Storage conditions:
    4 °C (long term), −20 °C (for standard)
  • Detection method:
    Colorimetric 450 nm
  • Sensitivity:
    6,6 pg/ml (Range 7,81 - 500 pg/ml)
  • Tests per kit:
    96 wells

Specifications

About this item

The IL-2 (human), EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in 3 hours.

  • Low dynamic range provides sensitive measurements
  • Validated in a wide variety of matrices
  • Ready to use reagents and pre-coated plates reduce errors

For the quantitative determination of human IL-2 in culture supernatants, plasma, and serum.

Delivery: Supplied with microtiter plate, standard, antibody, conjugate, assay buffer 28, TMB substrate, stop solution 2, wash buffer concentrate.